EMAIL THIS PAGE TO A FRIEND
Genetics
Paternal Genome Elimination in
PMID 28292917
Abstract
How sex is determined in insects is diverse and dynamic, and includes male heterogamety, female heterogamety, and haplodiploidy. In many insect lineages, sex determination is either completely unknown or poorly studied. We studied sex determination in Psocodea-a species-rich order of insects that includes parasitic lice, barklice, and booklice. We focus on a recently discovered species of