EMAIL THIS PAGE TO A FRIEND
Clinical epigenetics
LINE-1 is preferentially hypomethylated within adenomatous polyps in the presence of synchronous colorectal cancer.
PMID 28293326
Abstract
Conventional tubular adenomas are frequently detected in patients undergoing average risk screening colonoscopy and are over-represented in patients who will develop colorectal cancer (CRC). Whether features of adenomas could serve as predictors of synchronous CRC is not known. Here, we investigate whether global methylation markers, including LINE-1, differ within adenomas in patients with and without synchronous CRC. Colorectal tubular/tubulovillous adenomatous polyps in the absence (P group, Global methylation (5-mC) showed no differences in overall methylation status in the adenomatous polyps in the two groups (5-mC relative to control %, PC group 0.117; P group 0.161, LINE-1 hypomethylation of precancerous adenomas correlates with the presence of synchronous CRC. Measurement of DNA hypomethylation levels of colorectal adenomas by LINE-1 could have future implications in approaches to defining CRC risk in screening programs.