EMAIL THIS PAGE TO A FRIEND
Experimental physiology
A translational approach for NMDA receptor profiling as a vulnerability biomarker for depression and schizophrenia.
PMID 28294453
Abstract
What is the central question of this study? Can the change in plasma arginine vasopressin concentration (P