EMAIL THIS PAGE TO A FRIEND
Clinical and experimental immunology
Targeting C3a/C5a receptors inhibits human mesangial cell proliferation and alleviates immunoglobulin A nephropathy in mice.
PMID 28295247
Abstract
Complement activation has a deep pathogenic influence in immunoglobulin (Ig)A nephropathy (IgAN). C3a and C5a, small cleavage fragments generated by complement activation, are key mediators of inflammation. The fragments exert broad proinflammatory effects by binding to specific receptors (C3aR and C5aR, respectively). However, no studies thus far have investigated the effects of C3a, C5a and their receptors on IgAN. We observed that C3aR and C5aR antagonists repressed IgA-induced cell proliferation and interleukin (IL)-6 and monocyte chemotactic protein 1 (MCP-1) production in cultured human mesangial cells (HMCs). Furthermore, an IgAN mouse model induced by Sendai virus infection was employed to investigate the effects of C3aR and C5aR on IgAN in vivo for the first time. Wild-type (WT) and several knock-out mouse strains (C3aR
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1192
SB290157 trifluoroacetate salt, ≥97% (HPLC)
C22H28N4O4 · CF3CO2H