EMAIL THIS PAGE TO A FRIEND
Journal of thrombosis and haemostasis : JTH
Measurement of platelet aggregation, independently of patient platelet count: a flow-cytometric approach.
PMID 28296243
Abstract
Essentials Platelet function may influence bleeding risk in thrombocytopenia, but useful tests are needed. A flow cytometric platelet aggregation test independent of the patient platelet count was made. Platelet aggregation was reduced in thrombocytopenic patients with hematological cancer. High platelet aggregation ruled out bleeding tendency in thrombocytopenic patients. Background Methods for testing platelet aggregation in thrombocytopenia are lacking. Objective To establish a flow-cytometric test of in vitro platelet aggregation independently of the patient's platelet count, and examine the association of aggregation with a bleeding history in thrombocytopenic patients. Patients/methods We established a flow-cytometric assay of platelet aggregation, and measured samples from healthy individuals preincubated with antiplatelet drugs, and samples from two patients with inherited platelet disorders. Then, we included 19 healthy individuals and 20 patients with platelet counts of ≤ 50 × 10