EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Degradation of organic pollutants by bio-inspired rectangular and hexagonal titanium dioxide nanostructures.
PMID 28297682
Abstract
Dyes are used in textile, printing, leather, pharmaceutical, food and cosmetic industries. Dyes add color and pattern to materials. The presence of even very low concentration of dyes/dyes degradation products in effluent is highly toxic to humans and aquatic organisms. It is important to remove these dye degradation pollutants from the industrial effluents before their disposal. In recent years nanoparticles have been used for the removal of dyes from industrial waste water. Titanium dioxide nanostructures (TiO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
38800
Malachite Green chloride, analytical standard
C23H25ClN2