EMAIL THIS PAGE TO A FRIEND
BioTechniques
Antibody incubation at 37°C improves fluorescent immunolabeling in free-floating thick tissue sections.
PMID 28298178
Abstract
Fluorescent immunolabeling and imaging in free-floating thick (50-60 μm) tissue sections is relatively simple in practice and enables design-based non-biased stereology, or 3-D reconstruction and analysis. This method is widely used for 3-D in situ quantitative biology in many areas of biological research. However, the labeling quality and efficiency of standard protocols for fluorescent immunolabeling of these tissue sections are not always satisfactory. Here, we systematically evaluate the effects of raising the conventional antibody incubation temperatures (4°C or 21°C) to mammalian body temperature (37°C) in these protocols. Our modification significantly enhances the quality (labeling sensitivity, specificity, and homogeneity) and efficiency (antibody concentration and antibody incubation duration) of fluorescent immunolabeling of free-floating thick tissue sections.