EMAIL THIS PAGE TO A FRIEND
Reproduction, fertility, and development
Prostaglandin F
PMID 28298246
Abstract
The endometrium of domestic animals undergoes regular periods of regeneration and degeneration and exhibits a remarkable capacity for self-repair during the oestrous cycle. The endometrial growth pattern is also observed during in the implantation period and early pregnancy, but the mechanism underlying endometrial growth in these processes remains unclear. A positive correlation between endometrial growth in these processes and prostaglandin (PG) F
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8549
Fluprostenol, 10 mg/mL in ethanol, 98%
C23H29F3O6