EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering
Chlorine dioxide oxidation of Escherichia coli in water - A study of the disinfection kinetics and mechanism.
PMID 28301286
Abstract
This study investigated the kinetics and mechanism of chlorine dioxide (ClO
Related Materials