EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Respiration-Deficient Astrocytes Survive As Glycolytic Cells
PMID 28314814
Abstract
Neurons and glial cells exchange energy-rich metabolites and it has been suggested, originally based on
Related Materials