European journal of pharmacology
Inhibition of osteoporosis by the αvβ3 integrin antagonist of rhodostomin variants.
PMID 28315346
Integrins are heterodimeric cell surface receptors that mediate cell-cell and cell-matrix interaction. The vitronectin and osteopontin receptor αvβ3 integrin has increased expression levels and is implicated in the adhesion, activation, and migration of osteoclasts on the bone surface as well as osteoclast polarization. αvβ3 integrin plays an important role in osteoclast differentiation and resorption. In addition, Arg-Gly-Asp (RGD)-containing peptides, small molecular inhibitors, and antibodies to αvβ3 integrin have been shown to inhibit bone resorption in vitro and in vivo. Here we examined the effects of a disintegrin HSA-ARLDDL a genetically modified mutant of rhodostomin conjugated with human serum albumin, which is highly selective of αvβ3, on RANKL-induced osteoclastogenesis and ovariectomy (OVX)-induced osteoporosis. In RANKL-induced osteoclastogenesis, HSA-ARLDDL significantly inhibited osteoclast formation, and IC50 was at nM range. Post-treatment HSA-ARLDDL also inhibits osteoclast formation. Furthermore, weekly administration of HSA-ARLDDL significantly inhibits the increase in serum bone resorption marker levels and decrease in cancellous bone loss in tibia and femur induced by OVX. On the other hand, HSA-ARLDDL did not affect the differentiation and calcium deposition of osteoblasts. These results indicate that the highly selective and long-acting αvβ3 integrin antagonists could be developed as effective drugs for postmenopausal osteoporosis.