EMAIL THIS PAGE TO A FRIEND
Behavioural brain research
TAT-PEP, a novel blocker of PirB, enhances the recovery of cognitive function in mice after transient global cerebral ischemia.
PMID 28315758
Abstract
Neuronal damage and axonal regeneration inhibition are the main reasons to poor functional recovery after ischemia. Nogo-A signals inhibit axon outgrowth through the PirB receptor after ischemic reperfusion injury in central nervous system. We use TAT-PEP, a novel protein which could pass through the blood brain barrier, to block the function of PirB and identify the long-term neurological and behavioral recovery after bilateral common carotid artery occlusion (BCCAO) in mice. We observed that TAT-PEP promoted neuron survival and inhibited neuronal apoptosis. TAT-PEP increased the expression of Tau, GAP43 and MAP-2 proteins. In addition, the short-term and long-term cognitive functions were also enhanced, indicating that TAT-PEP had a long-term neuroprotective effect, which reduced neurologic injury and neuron loss, promoted neurite outgrowth and enhanced functional recovery after ischemia. These studies reveal the mechanism of PirB on stroke and offer a potential therapeutic method for cerebral ischemia in humans.