EMAIL THIS PAGE TO A FRIEND
Nanoscale
Albumin-based nanoparticles loaded with hydrophobic gadolinium chelates as T
PMID 28317976
Abstract
Magnetic resonance contrast agents with T
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
339652
Cyclen, 97%
C8H20N4