EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Mechanisms of toxic action of silver nanoparticles in the protozoan Tetrahymena thermophila: From gene expression to phenotypic events.
PMID 28318795
Abstract
Silver nanoparticles (AgNPs) are highly toxic to aquatic organisms, however, there is no consensus whether the toxicity is caused solely by released Ag-ions or also by reactive oxygen species (ROS). Here, the effects of protein-coated AgNPs (14.6 nm, Collargol) were studied on viability, oxidative stress and gene expression levels in wild type strains (CU427 and CU428) of ciliate Tetrahymena thermophila. Viability-based 24 h EC
Related Materials