EMAIL THIS PAGE TO A FRIEND
Stem cell research & therapy
Dysregulated systemic lymphocytes affect the balance of osteogenic/adipogenic differentiation of bone mesenchymal stem cells after local irradiation.
PMID 28320453
Abstract
While it is known that irradiation can induce local and systemic bone loss over time, how focal irradiation induces systemic bone complications remains unclear. Immune cells are thought to be crucial to bone homeostasis, and abnormal immune cells lead to serious disruption of bone homeostasis, such as in acute lymphoblastic leukaemia. This disruption primarily occurs due to inhibition of the osteogenic differentiation of bone mesenchymal stem cells (BMSCs). In this study, we detected local and systemic bone loss in trabecular bone by micro-computed tomography (micro-CT) and measurement of peroxisome proliferator-activated receptor gamma (PPARγ) and runt-related transcription factor 2 (RUNX2) expression in BMSCs using real-time polymerase chain reaction and western blotting. Additionally, changes in lymphocytes (B cells and CD4 Focal irradiation induced local and systemic bone loss in trabecular bone. Increased PPARγ expression and decreased RUNX2 expression were observed, accompanied by upregulated adipogenesis and downregulated osteogenesis of BMSCs. B cells and CD8 Focal irradiation induced local and systemic bone loss in trabecular bone. Increased B cells and CD8