EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
A simple synthetic route to polyoxovanadate-based organic-inorganic hybrids using EEDQ as an ester coupling agent.
PMID 28322395
Abstract
A reaction strategy for the post-functionalization of hexavanadate derivatives is presented herein. In this study, five polyoxovanadate-based organic-inorganic hybrids TBA
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D221309
1,12-Dodecanediol, 99%
C12H26O2