EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Monocyte activation drives preservation of membrane thiols by promoting release of oxidised membrane moieties via extracellular vesicles.
PMID 28323130
Abstract
The redox state of cellular exofacial molecules is reflected by the amount of available thiols. Furthermore, surface thiols can be considered as indicators of immune cell activation. One group of thiol containing proteins, peroxiredoxins, in particular, have been associated with inflammation. In this study, we assessed surface thiols of the U937 and Thp1 monocyte cell lines and primary monocytes in vitro upon inflammatory stimulation by irreversibly labelling the cells with a fluorescent derivative of maleimide. We also investigated exofacial thiols on circulating blood mononuclear cells in patients with rheumatoid arthritis and healthy controls. When analysing extracellular vesicles, we combined thiol labelling with the use of antibodies to specific CD markers to exclude extracellular vesicle mimicking signals from thiol containing protein aggregates. Furthermore, differential detergent lysis was applied to confirm the vesicular nature of the detected extracellular events in blood plasma. We found an increase in exofacial thiols on monocytes upon in vitro stimulation by LPS or TNF, both in primary monocytes and monocytic cell lines (p<0.0005). At the same time, newly released extracellular vesicles showed a decrease in their exofacial thiols compared with those from unstimulated cells (p<0.05). We also found a significant elevation of surface thiols on circulating monocytes in rheumatoid arthritis patients (p<0.05) and newly released extracellular vesicles of isolated CD14