EMAIL THIS PAGE TO A FRIEND
Head & neck
Induction of tumor regression by intratumoral STING agonists combined with anti-programmed death-L1 blocking antibody in a preclinical squamous cell carcinoma model.
PMID 28323387
Abstract
Cyclic dinucleotides (CDNs) are bacterial intracellular messengers that have demonstrated antitumor activity in melanoma and breast tumors, although their role in immunotherapy of head and neck squamous cell cancers (HNSCCs) has not been well investigated. We measured primary tumor growth rates, mechanism of antitumor activity, and efficacy of programmed death-L1 blockade combinatorial therapy in SCCFVII tumor-bearing C3H/HeOUJ mice undergoing intratumoral injections with RR-cyclic-di-guanine (synthetic CDG), CDG (natural cyclic-di-guanine), R848 (TLR 7/8 agonist), or phosphate buffered saline (PBS, control). Intratumoral CDN treatment groups showed decreased tumor size and enhanced splenocyte Th1 response when compared to the PBS treatment control group (p < .05). The RR-CDG tumor microenvironment showed upregulated interferon (IFN)-γ+CD8+ and programmed death-L1. Combining programmed death-L1 blocking antibody with RR-CDG induced regression of established tumors. This study demonstrates the antitumor effects of CDNs in a HNSCC cell line. These preclinical data strongly support the future clinical development of intratumoral CDN in patients with HNSCC. © 2017 Wiley Periodicals, Inc. Head Neck 39: 1086-1094, 2017.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0196
Resiquimod, ≥98% (HPLC)
C17H22N4O2