EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Regulation of secretion and enzymatic activity of lipoprotein lipase by C-mannosylation.
PMID 28327359
Abstract
Lipoprotein lipase (LPL) is a crucial enzyme in lipid metabolism and transport, and its enzymatic deficiency causes metabolic disorders, such as hypertriglyceridemia. LPL has one predicted C-mannosylation site at Trp