EMAIL THIS PAGE TO A FRIEND
3 Biotech
In vitro cytotoxicity and antibacterial activity of silver-coated electrospun polycaprolactone/gelatine nanofibrous scaffolds.
PMID 28330282
Abstract
The development of nano-sized scaffolds with antibacterial properties that mimic the architecture of tissue is one of the challenges in tissue engineering. In this study, polycaprolactone (PCL) and PCL/gelatine (Ge) (70:30) nanofibrous scaffolds were fabricated using a less toxic and common solvent, formic acid and an electrospinning technique. Nanofibrous scaffolds were coated with silver (Ag) in different concentrations of silver nitrate (AgNO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3813 Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions