EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Chlorinated pesticides and natural brominated anisoles in air at three northern Baltic stations.
PMID 28336095
Abstract
Polyurethane foam (PUF) disk passive samplers were deployed at one inland and two island locations in the Bothnian Bay region of the northern Baltic Sea. Uptake was linear over 81-147 d and a temperature range of -2.6 to 14.2 °C for organochlorine pesticides (OCPs) and current-use pesticides (CUPs) having log K
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
360953
α,α-Dicyanoethyl acetate, 98%
C6H6N2O2