EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Pharmacological assessments of potent A
PMID 28336342
Abstract
A
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2387
5′-(N-Ethylcarboxamido)adenosine, powder
C12H16N6O4