EMAIL THIS PAGE TO A FRIEND
Plant physiology
GmILPA1, Encoding an APC8-like Protein, Controls Leaf Petiole Angle in Soybean.
PMID 28336772
Abstract
Leaf petiole angle (LPA) is an important plant architectural trait that affects canopy coverage, photosynthetic efficiency, and ultimately productivity in many legume crops. However, the genetic basis underlying this trait remains unclear. Here, we report the identification, isolation, and functional characterization of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E8640
E-64d, protease inhibitor
C17H30N2O5
P3683 Paraplast Plus®, for tissue embedding