EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
A column switching ultrahigh-performance liquid chromatography-tandem mass spectrometry method to determine anandamide and 2-arachidonoylglycerol in plasma samples.
PMID 28337517
Abstract
This study reports a fast, sensitive, and selective column switching ultrahigh-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) method to determine the endocannabinoids (eCBs), anandamide (AEA), and 2-arachidonoylglycerol (2-AG) in plasma samples. This bidimensional system used a restricted access media column (RP-8 ADS, 25 mm × 4 mm × 25 μM) in the first dimension and a core-shell Kinetex C18 (100 mm × 2, 1.7 mm × 1 μM) column in the second dimension, followed by detection in a mass spectrometer triple quadrupole (multiple reactions monitoring mode) operating in the positive mode. RP-8 ADS was used for trace enrichment of eCBs (reverse phase partitioning) and macromolecular matrix size exclusion; the core-shell column was used for the chromatographic separation. The column switching UHPLC-MS/MS method presented a linear range spanning from 0.1 ng mL