EMAIL THIS PAGE TO A FRIEND
Acta oto-laryngologica
The modified temporalis muscle flap in reconstruction of palate and temporal deformity.
PMID 28338372
Abstract
The purpose of this article was to introduce a modified temporalis muscle flap (TMF) which was used to reconstruct palate and temporal deformity. This was a retrospective review of the use of the modified TMF in reconstruction of palate and temporal deformity. We evaluated the result which included operative time, bleeding, necrosis, infection, facial nerve deficit, and cosmetic deformity. All the 16 patients accepted the modified TMF surgery successfully. There were no complications. All of them were satisfied with the postoperative appearance. The modified TMF was a reliable and safe flap that can be used to reconstruct the surgical defect of plate and fill the temporal fossa.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4080
6, 2′, 4′-trimethoxyflavone, ≥98% (HPLC)
C18H16O5