EMAIL THIS PAGE TO A FRIEND
PMID 28338421
Abstract
Advanced biomaterials that are capable of guiding robust bone regeneration are highly demanded for translational therapy of bone defects or bone augmentation in clinics. One of the strategic approaches is to produce tissue engineering (TE) constructs that mediate bone regeneration by recapitulating the natural bone formation or healing process. In this study, we aimed at producing devitalized mineralized carriers with augmented bone forming capacity via a modified culture protocol (i.e., culture conditions with high calcium and/or phosphate concentrations) that first promotes cell growth and, subsequently, mineralized extracellular matrix (ECM) deposition by human periosteum-derived osteoprogenitor cells (hPDCs) on additive manufactured three-dimensional (3D) porous titanium (Ti)-based scaffolds. Qualitative and quantitative analysis was performed to characterize the physicochemical properties of the produced devitalized mineralized carriers, as well as their effects as carriers on in vitro cell growth and osteochondrogenic differentiation of hPDCs under a perfusion bioreactor culture set-up. The results showed that the modified culture protocol was useful to produce devitalized mineralized carriers with different amount, distribution, composition, and morphology of mineralized matrix that resembled hydroxyapatite, and exhibited different Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
469548
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
C8H11F3O2