EMAIL THIS PAGE TO A FRIEND
Journal of environmental management
Evaluation of heavy crude oil from a water-oil model system as starting material for the preparation of adsorbents type NaY zeolite-templated carbon.
PMID 28342341
Abstract
In this work, NaY zeolite is explored as a possible "template" to obtain porous materials type ZTC from the adsorption of heavy crude oil in a water-oil model system (emulsion). In order to produce the adsorbents, a cationic surfactant is selected to facilitate the adsorption of the crude oil into the pores of the zeolite and to get the composite, which was activated with controlled thermal treatments (T: 700-800 °C and t: 0.5-1 h) in inert conditions (N
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1800900
Methylthioninium chloride, European Pharmacopoeia (EP) Reference Standard
C16H18ClN3S · xH2O