EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
The effects of propofol on the growth behavior of hepatoma xenografts in Balb/c mice.
PMID 28342365
Abstract
Studies on the effects of propofol on the growth of hepatoma xenografts in Balb/c mice. In an effort to establish a hepatoma-xenograft model of BALB/C mice, human hepatocellular carcinoma cells SMMC-7721 were inoculated subcutaneously into BALB/C mice. Forty mice were randomly divided into five different groups (n=8): control group (C group), Intralipid group (Y group), low dose (50mg/kg) propofol group (P No statistical significance in the tumor volume at T Within a certain range, propofol was found to inhibit tumor growth and expression of MMP-2 and VEGF proteins in hepatoma xenografts in BALB/C mice in a dose-dependent manner.