EMAIL THIS PAGE TO A FRIEND
Neuropeptides
The effects of endothelin-1 on satellite glial cells in peripheral ganglia.
PMID 28342550
Abstract
Endothelins (ET) are a family of highly active neuropeptides with manifold influences via ET receptors (ETR) in both the peripheral and central nervous systems. We have shown previously that satellite glial cells (SGCs) in mouse trigeminal ganglia (TG) are extremely sensitive to ET-1 in evoking [Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B150
BQ-123, ≥99%, sodium salt, lyophilized powder
C31H41N6O7 Na