EMAIL THIS PAGE TO A FRIEND
Meat science
The effect of the packaging system and storage time on myofibrillar protein degradation and oxidation process in relation to beef tenderness.
PMID 28342838
Abstract
This study investigated the impact of packaging systems on the degradation and oxidation of beef proteins regarding beef tenderness of longissimus lumborum (LL) and biceps femoris (BF) muscles stored in vacuum skin packaging (VSP), a modified atmosphere with high oxygen concentration (MAP), and combined of these two methods (VSP+MAP). A significant decrease in the Warner-Bratzler shear force (WBSF) in VSP at D14 and D28 for LL was observed compared to BF. A significant effect of packaging system on troponin-T (Tn-T) and desmin degradation was shown (p≤0.001). A high concentration of oxygen in MAP and VSP+MAP affected protein oxidation, which was reflected in myosin oxidative cross-linking. An increase of WBSF values detected in steaks packed in VSP and VSP+MAP systems could be caused by the intensification of protein oxidation. Furthermore, BF was more susceptible to oxidation compared to LL. The VSP+MAP packaging system has resulted in the maintenance of a bright, red color, however has not improved the beef tenderness.
Related Materials