EMAIL THIS PAGE TO A FRIEND
Irish veterinary journal
Application of antibodies to recombinant heat shock protein 70 in immunohistochemical diagnosis of
PMID 28344769
Abstract
Detection of MAP Heat shock protein 70 gene was amplified and cloned into an expression vector, Champion pET-SUMO, then expressed in Anti-HSP70 produced in this study offers an opportunity for improved diagnosis, screening of MAP in animal tissues and in studies on the pathogenesis of MAP.