EMAIL THIS PAGE TO A FRIEND
SLAS discovery : advancing life sciences R & D
A High-Throughput Method for Measuring Drug Residence Time Using the Transcreener ADP Assay.
PMID 28346107
Abstract
Analysis of drug-target residence times during drug development can result in improved efficacy, increased therapeutic window, and reduced side effects. Residence time can be estimated as the reciprocal of the dissociation rate ( k