EMAIL THIS PAGE TO A FRIEND
BMC biology
TriPer, an optical probe tuned to the endoplasmic reticulum tracks changes in luminal H
PMID 28347335
Abstract
The fate of hydrogen peroxide (H Here we report on the development of TriPer, a vital optical probe sensitive to changes in the concentration of H A tri-cysteine system with a single peroxidatic thiol enables H