EMAIL THIS PAGE TO A FRIEND
Protein expression and purification
Expression, purification and characterization of active untagged recombinant human leukemia inhibitory factor from E.coli.
PMID 28347752
Abstract
Leukemia inhibitory factor (LIF), a member of the IL-6 cytokine family, is considered to be a pleiotropic cytokine and functions in both cell proliferation and differentiation. It is widely used in the culture of mouse embryonic stem cells and is implicated in the implantation of mouse model and possibly in humans. Great efforts have been made on the efficient generation of LIF to meet the requirement of this cytokine in biomedical research. However, because of the low expression level in the eukaryotic system and poor purification yields, recombinant human LIF has usually been expressed either as inclusion body or as fusion protein in E. coli (Escherichia coli). Here we introduce a simple method to express hLIF in a soluble form in E. coli and a subsequent purification method. The expression of hLIF was induced at a low temperature (16 °C) and most of the expressed hLIF was observed to be in a soluble form. Then by using three steps of chromatography, which could be easily scaled-up for industrial purposes, active untagged hLIF was purified with similar bioactivity compared to that of the commercial product. The endotoxin level of purified hLIF protein in our method was determined to be lower than 1EU/μg, which was also comparable to the commercial products. Furthermore, as hLIF was expressed in a soluble form, there was no need to develop the denaturation and renaturation methods. The yield of hLIF protein was evaluated to be approximately 0.7 mg hLIF from 1 g wet weight of E.coli in our method.