EMAIL THIS PAGE TO A FRIEND
Journal of biotechnology
Biosynthesis of an antiviral compound using a stabilized phosphopentomutase by multipoint covalent immobilization.
PMID 28347766
Abstract
Ribavirin is a synthetic guanosine analogue with a broad-spectrum of antiviral activity. It is clinically effective against several viruses, such as respiratory syncytial virus, several hemorrhagic fever viruses and HCV when combined with pegylated interferon-α. Phosphopentomutase (PPM) catalyzes the transfer of intramolecular phosphate (from C1 to C5) on ribose, and is involved in pentose phosphate pathway and in purine metabolism. Reactions catalyzed by this enzyme are useful for nucleoside analogues production. However, out of its natural environment PPM is unstable and its stability is affected by parameters such as pH and temperature. Therefore, to irreversibly immobilize this enzyme, it needs to be stabilized. In this work, PPM from Escherichia coli ATCC 4157 was overexpressed, purified, stabilized at alkaline pH and immobilized on several supports. The activity of different additives as stabilizing agents was evaluated, and the best result was found using 10% (v/v) glycerol. Under this condition, PPM maintained 86% of its initial activity at pH 10 after 18h incubation, which allowed further covalent immobilization of this enzyme on glyoxyl-agarose with a high yield. This is the first time that PPM has been immobilized by multipoint covalent attachment on glyoxyl support, this derivative being able to biosynthesize ribavirin from α-d-ribose-5-phosphate.