EMAIL THIS PAGE TO A FRIEND
The Journal of experimental biology
Olfactory sensitivity of the marine flatfish
PMID 28348043
Abstract
Chemical communication is better understood in freshwater fish than marine fish. The Senegalese sole (