EMAIL THIS PAGE TO A FRIEND
The Libyan journal of medicine
How does serum brain natriuretic peptide level change under nasal continuous positive airway pressure in obstructive sleep apnea-hypopnea syndrome?
PMID 28349847
Abstract
Background Obstructive sleep apnea-hypopnea syndrome (OSAHS) is associated with cardiovascular morbidity and mortality, which can be improved by using continuous positive airway pressure (CPAP) therapy. However, the pathophysiological links between the two kinds of disease and the mechanism of the CPAP effect remain incompletely understood. We aimed to inquire into the myocardial involvement in this relationship. We suggested that serum brain natriuretic peptide (BNP) is sensitive enough to detect myocardial stress caused by OSAHS. Design and methods Sixty-four subjects without cardiovascular disease (21 controls, 24 normotensive OSAHS patients, and 19 hypertensive OSAHS patients) were analyzed for serum BNP at baseline and serially over 6 months. CPAP was applied to 23 patients with severe OSAHS. Results At baseline, the serum BNP levels were significantly higher (p=0.0001) in the OSAHS group (22.3±14.79 pg/ml) than in the control group (9.2±6.75 pg/ml). Increased serum BNP levels were significantly associated with mean transcutaneous oxygen saturation (SpO