EMAIL THIS PAGE TO A FRIEND
Scientific reports
A new fluorescent probe for colorimetric and ratiometric detection of sulfur dioxide derivatives in liver cancer cells.
PMID 28349998
Abstract
A new ratiometric fluorescent probe was constructed with hemicyanine and 7-nitrobenzofurazan for detection of sulfur dioxide derivatives (HSO