EMAIL THIS PAGE TO A FRIEND
Frontiers in molecular neuroscience
Myosin IIA-related Actomyosin Contractility Mediates Oxidative Stress-induced Neuronal Apoptosis.
PMID 28352215
Abstract
Oxidative stress-induced neuronal apoptosis plays an important role in the progression of central nervous system (CNS) diseases. In our study, when neuronal cells were exposed to hydrogen peroxide (H