EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Flexible organofunctional aerogels.
PMID 28352868
Abstract
Flexible inorganic-organic silica aerogels based on methyltrimethoxysilane (MTMS, CH