EMAIL THIS PAGE TO A FRIEND
Oncology letters
MicroRNA-149 targets specificity protein 1 to suppress human tongue squamous cell carcinoma cell proliferation and motility.
PMID 28356969
Abstract
The expression and function of microRNA-149 have been studied in numerous types of cancer. However, thus far, there are no studies of microRNA-149 in tongue squamous cell carcinoma (TSCC). The present study investigated the expression, biological function and molecular mechanism of microRNA-149 in TSCC