EMAIL THIS PAGE TO A FRIEND
Oncology letters
Clinical significance of platelet-derived growth factor receptor-β gene expression in stage II/III gastric cancer with S-1 adjuvant chemotherapy.
PMID 28356977
Abstract
Overall survival remains unsatisfactory in stage II/III gastric cancer, even after curative surgery and adjuvant chemotherapy. Platelet-derived growth factor receptor-β (