EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A bidirectional synthesis of spiroacetals via Rh(ii)-catalysed C-H insertion.
PMID 28357432
Abstract
Acyclic methylene acetals bearing two diazoester subunits have been converted to [5,5]-spiroacetals via bidirectional C-H insertion under Rh(ii) catalysis. Using a chiral Rh(ii) catalyst, the major diastereomer can be produced in high enantiomeric excess (89%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
905259
Rh2(S-PTAD)4
C80H80N4O16Rh2