EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Oncostatin M in William's E medium is suitable for initiation of hepatocyte differentiation in human induced pluripotent stem cells.
PMID 28358419
Abstract
William's E (WE) is a suitable medium for the differentiation of human induced pluripotent stem (iPS) cells to the hepatocyte lineage. The aim of the present study was to investigate various growth factors in their ability to promote hepatocyte differentiation of iPS cells in WE medium. Human iPS 201B7 cells were cultured in WE medium supplemented with growth factors, and mRNA expression levels and promoter activities of α‑fetoprotein (AFP) and albumin were examined by reverse transcription‑quantitative polymerase chain reaction and luciferase assay, respectively. In addition, time course analysis of AFP mRNA expression was performed in 201B7 cells cultured in WE medium supplemented with oncostatin M. The results demonstrated that mRNA expression levels of AFP were significantly elevated by most growth factors tested as supplements in WE medium, except all‑trans retinoic acid, compared with cells cultured in ReproFF (a medium that maintains pluripotency). The highest increase in AFP mRNA expression levels was observed by oncostatin M stimulation. Albumin mRNA expression levels were increased by all‑trans retinoic acid and insulin‑transferrin‑selenium supplementation in WE medium compared with cells cultured in ReproFF. Oncostatin M supplementation significantly stimulated the promoter activity of the AFP gene, but no growth factor tested significantly stimulated the promoter activity of the albumin gene. By time course analysis, significant increase of AFP mRNA expression was observed on the sixth day post‑stimulation, compared with cells cultured in WE medium alone. In conclusion, the present study demonstrated that oncostatin M supplementation in WE medium was sufficient to initiate hepatocyte differentiation in iPS cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R2625
Retinoic acid, ≥98% (HPLC), powder
C20H28O2