EMAIL THIS PAGE TO A FRIEND
Molecular cancer research : MCR
Insulin Receptor and GPCR Crosstalk Stimulates YAP via PI3K and PKD in Pancreatic Cancer Cells.
PMID 28360038
Abstract
We examined the impact of crosstalk between the insulin receptor and G protein-coupled receptor (GPCR) signaling pathways on the regulation of Yes-associated protein (YAP) localization, phosphorylation, and transcriptional activity in the context of human pancreatic ductal adenocarcinoma (PDAC). Stimulation of PANC-1 or MiaPaCa-2 cells with insulin and neurotensin, a potent mitogenic combination of agonists for these cells, promoted striking YAP nuclear localization and decreased YAP phosphorylation at Ser
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0525
kb-NB142-70, ≥97% (HPLC)
C11H9NO2S2
SML0382
KU 0063794, ≥98% (HPLC)
C25H31N5O4
N6383
Neurotensin, ≥90% (HPLC)
C78H121N21O20