EMAIL THIS PAGE TO A FRIEND
Development (Cambridge, England)
Numb regulates somatic cell lineage commitment during early gonadogenesis in mice.
PMID 28360133
Abstract
During early gonadogenesis, proliferating cells in the coelomic epithelium (CE) give rise to most of the somatic cells in both XX and XY gonads. Previous dye-labeling experiments showed that a single CE cell could give rise to additional CE cells and to both supporting and interstitial cell lineages, implying that cells in the CE domain are multipotent progenitors, and suggesting that an asymmetric division is involved in the acquisition of gonadal cell fates. We found that NUMB is asymmetrically localized in CE cells, suggesting that it might be involved. To test this hypothesis, we conditionally deleted
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5942
DAPT, ≥98% (HPLC), solid
C23H26F2N2O4