EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Polyadenylate polymerase in the diagnosis of the early stages of chemical carcinogenesis].
PMID 2836273
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31534
2-Imidazolidone, PESTANAL®, analytical standard
C3H6N2O
I601
2-Imidazolidone, 96%
C3H6N2O