EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Retinoic Acid Regulates Immune Responses by Promoting IL-22 and Modulating S100 Proteins in Viral Hepatitis.
PMID 28363907
Abstract
Although large amounts of vitamin A and its metabolite all-
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU125681 MISSION® esiRNA, esiRNA human S100A4 (esiRNA1)