EMAIL THIS PAGE TO A FRIEND
Scientific reports
Local delivery of novel MRTF/SRF inhibitors prevents scar tissue formation in a preclinical model of fibrosis.
PMID 28364121
Abstract
The myocardin-related transcription factor/serum response factor (MRTF/SRF) pathway represents a promising therapeutic target to prevent fibrosis. We have tested the effects of new pharmacological inhibitors of MRTF/SRF signalling in a preclinical model of fibrosis. CCG-222740, a novel MRTF/SRF inhibitor, markedly decreased SRF reporter gene activity and showed a greater inhibitory effect on MRTF/SRF target genes than the previously described MRTF-A inhibitor CCG-203971. CCG-222740 was also five times more potent, with an IC