EMAIL THIS PAGE TO A FRIEND
International orthopaedics
Exercise affects biological characteristics of mesenchymal stromal cells derived from bone marrow and adipose tissue.
PMID 28364139
Abstract
Both bone marrow mesenchymal stromal cells (BMSCs) and adipose-derived mesenchymal stromal cells (ADSCs) are good sources for tissue engineering. To maximize therapeutic efficacy of MSCs, an appropriate source of MSCs should be selected according to their own inherent characteristics for future clinical application. Hence, this study was conducted to compare proliferative, differential and antiapoptosis abilities of both MSCs derived from exercised and sedentary rats under normal and hypoxia/serum deprivation conditions (H/SD). Our results showed that exercise may enhance proliferative ability and decrease adipogenic ability of BMSCs and ADSCs. However, positive effect of exercise on osteogenesis was only observed for BMSCs in either environment. Little effect was observed on the antiapoptotic ability of both MSC types. It was also suggested that biological characteristics of both types were partly changed. It is therefore believed that BMSCs derived from exercised rat on early passage may be a good cell source for bone tissue engineering.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0332
Edelfosine, ≥95% (HPLC)
C27H58NO6P